Learn More
Eisco™ Essential Oils Extraction Apparatus
SCI_ED
Catalog No. S83778 Shop All Eisco Products
Change view
Click to view available options
Material:
Borosilicate Glass 3.3
Catalog No. Material
S83778 Borosilicate Glass 3.3
1 options
Catalog No. S83778 Supplier Eisco™ Supplier No. CH1050
Add to Cart
Add to Cart
The Essential Oils Extraction Apparatus includes everything needed to extract essential oils from plant material using glass steam distillation.
  • 23 piece essential oils extraction kit is made of high quality borosilicate 3.3 glass
  • Complete set up for extracting essential oils using glass steam distillation
  • Low temperature gradient
  • Superior chemical durability
  • Autoclavable
Includes
  • (2) 125 mL Pear Shaped Separating Funnels - PTFE Stopcock and Stem with Cone, (2) 500 mL Boiling Flasks - Round Bottom, (1) 500 mL Biomass Flask, (1) Multiple Adapter - Two Parallel Necks, (1) Vertical Recovery Bend Adapter - with Thermometer Socket, (1) Receiver Adapter, (1) Jointed Liebig Condenser, (10) Green Joint Clips - Size 24, (2) Polypropylene 24/40 Interchangeable Stoppers, (1) Soft Rubber Tubing, (1) Cork Flask Stand
Specifications
Includes (2) 125 mL Pear Shaped Separating Funnels - PTFE Stopcock and Stem with Cone, (2) 500 mL Boiling Flasks - Round Bottom, (1) 500 mL Biomass Flask, (1) Multiple Adapter - Two Parallel Necks, (1) Vertical Recovery Bend Adapter - with Thermometer Socket, (1) Receiver Adapter, (1) Jointed Liebig Condenser, (10) Green Joint Clips - Size 24, (2) Polypropylene 24/40 Interchangeable Stoppers, (1) Soft Rubber Tubing, (1) Cork Flask Stand
For Use With (Application) Extracting Essential Oils Using Glass Steam Distillation
Type Distillation Apparatus
Material Borosilicate Glass 3.3
Quantity 23 Pk.
Autoclavable Autoclavable