Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Esterification Reagent, Reagents
Esterification Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CE1000001C
Quantity | 1 L |
---|
Chemical Identifiers
110-86-1, 85-44-9 | |
79.10 | |
pyridine |
C5H5N | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
C1=CC=NC=C1 |
Specifications
Colorless to Slight Yellow | |
C5H5N | |
C1=CC=NC=C1 | |
79.10 | |
Glass Bottle | |
1 L | |
Liquid |
110-86-1, 85-44-9 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
Reagent | |
1.0353 | |
Non-sterile | |
Esterification Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction