Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estradiol-d4 | 66789-03-5 | 99.6% | 276.41 | C18H20D4O2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB0141S110MG

Encompass_Preferred

Deuterium-labeled estradiol supplied as a solid for use as a tracer and internal standard in quantitative analyses by NMR, GC-MS, and LC-MS. Supplied in a 10 mg package with specified storage conditions for powder and solutions.

  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS analyses.
  • Chemical purity 99.55% and isotopic labeling for reliable quantitation.
  • Chemical formula C18H20D4O2; molecular weight 276.41.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C for long term; in solvent -80°C for extended storage.

Catalog No. 50-002-97199


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.