Learn More
Arbor Assays Estradiol ELISA Kit (5 Plate)
SDP
Supplier:  Arbor Assays K030H5
Encompass_Preferred
The DetectX Estradiol ELISA Kits use a specifically generated antibody to measure estradiol and its metabolites in urine and fecal samples. This kit is not recommended for serum, plasma, or saliva samples as the concentration of estradiol in these samples is too low to be measured without significant concentration.
Catalog No. 50-227-8431