Learn More
Alpco Ltd Estradiol ELISA
SDP
Supplier:  Alpco Ltd 11ESTHUE01
Encompass_Preferred
The Estradiol ELISA is for the direct quantitative determination of Estradiol by enzyme immunoassay in human Serum. Estradiol is one of the main components of naturally occurring estrogens and is the major estrogen secreted during the menstrual cycle. For ¿In Vitro Diagnostic¿ use within the United States of America. This product is for ¿Research Use Only¿ outside of the United States ofAmerica.
Catalog No. NC2299701
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.