Learn More
Arbor Assays Estradiol ELISA Kit (1 Plate)
SDP
Supplier:  Arbor Assays K030H1
Encompass_Preferred
The DetectX Estradiol ELISA Kits use a specifically generated antibody to measure estradiol and its metabolites in urine and fecal samples. This kit is not recommended for serum, plasma, or saliva samples as the concentration of estradiol in these samples is too low to be measured without significant concentration.
Catalog No. 50-227-8430
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.