Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Estradiol Parameter Assay Kit
SDP

Catalog No. KGE014 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KGE014 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. KGE014 Supplier R&D Systems™ Supplier No. KGE014
Add to Cart
Add to Cart
This item is not returnable. View return policy

Estradiol Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Estradiol Parameter Assay Kit quantifies multi-species Estradiol in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).

Specifications

Assay Range 12.30 - 3000pg/mL
Assay Sensitivity 12.1pg/mL
Conjugate Biotin
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Estradiol levels in various samples. This assay cross-reacts 0.07% with 17 alpha-ethynylestradiol, 0.03% with 17 beta-estradiol 3-Benzoate, 0.86% with Estriol, 0.26% with Estrone and 0.06% with Progesterone.
For Use With (Application) ELISA
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.