Learn More
Apexbio Technology LLC STING-IN-2 346691-38-1 25mg
SDP
Supplier:  Apexbio Technology LLC A468225
Encompass_Preferred
STING-IN-2 (CAS 346691-38-1) is a covalent small-molecule inhibitor targeting the stimulator of interferon genes (STING) protein in both murine and human systems By specifically inhibiting the STING signaling pathway STING-IN-2 reduces the activation of downstream immune and inflammatory responses Given the pivotal involvement of STING-mediated signaling in the pathogenesis of various autoimmune and inflammatory disorders STING-IN-2 serves as a specialized tool for investigating disease mechanisms and validating therapeutic strategies in preclinical models of autoimmunity and inflammation
Catalog No. 50-000-94987
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.