Learn More
Apexbio Technology LLC Alfuzosin HCl 81403-68-1 25mg
SDP
Supplier:  Apexbio Technology LLC A517325
Encompass_Preferred
Alfuzosin HCl (CAS 81403-68-1) is a functionally uroselective antagonist of sub 1 /sub adrenoceptors that exhibits no subtype selectivity among sub 1 /sub receptor isoforms By blocking sub 1 /sub adrenoceptors it inhibits smooth muscle contraction in the lower urinary tract In pharmacological assays Alfuzosin HCl suppresses phenylephrine-induced increases in intraurethral pressure by approximately 81% demonstrating significant activity with minimal cardiovascular effects This compound is utilized in research investigating sub 1 /sub adrenoceptor-mediated pathways relevant to urinary tract function and related pharmacology
Catalog No. 50-000-93791
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.