Learn More
Invitrogen™ Estrogen Receptor Beta Monoclonal Antibody (PPZ0506)
Shop All Thermo Scientific Products

Description
This antibody specifically recognizes human ER beta but does not recognize human ER alpha.
Specifications
Specifications
| Antigen | Estrogen Receptor Beta |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | PPZ0506 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Physiological saline with 0.1% sodium azide |
| Gene | ESR2 |
| Gene Accession No. | Q92731 |
| Gene Alias | ER; ER beta; ER[b]; Erb; Erb2; Erbeta; ER-beta; Esr2; ESRB; ESR-BETA; ESTRB; estrogen receptor 2; estrogen receptor 2 (beta); estrogen receptor 2 (ER beta); estrogen receptor 2 beta; estrogen receptor beta; estrogen receptor beta 1 isoform deletion variant; estrogen receptor beta 4; NR3A2; Nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta; steroid receptor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.