Learn More
Diagnostic Biosystems ESTROGEN RECEPTOR ER Estrogen Receptor (ER) - SP1
Supplier:  Diagnostic Biosystems RMAB001
Encompass_Preferred
This antibody recognizes a protein of 67kDa, which is identified as estrogen receptor (ER). ER content of breast cancer tissue is an important parameter in the prediction of prognosis and response to endocrine therapy. This antibody strongly stains the nucleus of epithelial cells in breast carcinomas. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.
Catalog No. 50-238-5696
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.