Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrone, 99+%, Thermo Scientific Chemicals

Catalog No. AC117840250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117840250 25 g Glass bottle
AC117840010 1 g Glass bottle
AC117840050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC117840250 Supplier Thermo Scientific Chemicals Supplier No. 117840250
  • $568.00
    $482.65 / Each of 1

    Save $85.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Estrone, 99%, CAS#53-16-7, is one of the major mammalian estrogen, is a steroid and a sex hormone. It is synthetized from cholesterol and secreted mainly from the gonads. It also presents antineoplastic and bone density conservative activities.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Estrone is an aromatized C18 steroid considered one of the major mammalian estrogen and a sex hormone. It shows antineoplastic and bone density conservative activities.

• This compound can interact with estrogen receptors in target tissues to produce similar effects to estradiol. Hormone-bound estrogen receptors dimerize, translocate to the nucleus of cells and bind to estrogen response elements (ERE) of genes. Binding to ERE alters the transcription rate of affected genes.

Applications

• Estrone has the ability to increase the hepatic synthesis of sex hormone binding globulin (SHBG), thyroid-binding globulin (TBG), and other serum proteins.

• It can suppress the release of follicle-stimulating hormone and luteinizing hormone.

• This compound can be produced from androstenedione or testosterone via estradiol. In vivo, it can be generated mainly in the ovaries, placenta, and peripheral tissues (especially adipose tissue) through conversion of androstenedione. It may be metabolized to 16-α-hydroxyestrone, which may be reduced to estriol by estradiol dehydrogenase.

• It has been used as medium supplement for hormone-based degranulation studies of natural killer cells.

• It is used as medium component for monitoring fatty acid synthase activity in breast adenocarcinoma cell lines.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-16-7
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.36
InChI Key DNXHEGUUPJUMQT-CBZIJGRNSA-N
Synonym estrone, folliculin, oestrone, theelin, estrovarin, estrugenone, follicular hormone, kestrone, estron, follicunodis
PubChem CID 5870
ChEBI CHEBI:17263
IUPAC Name (8R,9S,13S,14S)-3-hydroxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one
SMILES CC12CCC3C(C1CCC2=O)CCC4=C3C=CC(=C4)O

Specifications

Melting Point 258.0°C to 261.0°C
Color White
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (on dried substance) (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15,3763
Specific Rotation Condition + 158.00 (20.00°C c=1,dioxane)
Specific Rotation + 158.00
Solubility Information Solubility in water: 0.03g/L.
Formula Weight 270.36
Percent Purity 99+%
Clarity passes test
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Estrone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Suspected of causing cancer.
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-164-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.