Learn More
Etching Solution for Stainless Steel, Reagents
Etching Solution for Stainless Steel, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CE2375004A
Specifications
| Etching Solution for Stainless Steel | |
| 7647-01-0 , 10025-77-1 | |
| Reagent | |
| 1.3258 | |
| 4 L | |
| Liquid |
| Brown/Yellow | |
| 100°C | |
| Poly Bottle | |
| <1 | |
| Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.