Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva ETFE Tubing 2 m i.d

Catalog No. 502748590 Shop All Cytiva Products
Encompass

ETFE Tubing, 2 m, i.d. 0.25 mm, o.d. 1/16"

Catalog No. 50-274-8590 Supplier Cytiva Supplier No. 18112136
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ETFE Tubing, 2 m, i.d. 0.25 mm, o.d. 1/16"

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.