Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva ETFE Tubing Kit 10 x 1.0 m i.d

Catalog No. 502748584 Shop All Cytiva Products
Encompass

ETFE Tubing Kit 10 x 1.0 m, i.d. 1.0 mm, o.d. 1/16"

Catalog No. 50-274-8584 Supplier Cytiva Supplier No. 28980995
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ETFE Tubing Kit 10 x 1.0 m, i.d. 1.0 mm, o.d. 1/16"

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.