Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. E0035250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0035250G 250 g
E003525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0035250G Supplier TCI America Supplier No. E0035250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 594-44-5
Molecular Formula C2H5ClO2S
Molecular Weight (g/mol) 128.57
MDL Number MFCD00007460
InChI Key FRYHCSODNHYDPU-UHFFFAOYSA-N
Synonym ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride
PubChem CID 11667
IUPAC Name ethanesulfonyl chloride
SMILES CCS(Cl)(=O)=O

Specifications

Color Yellow
Boiling Point 171°C
Quantity 250 g
UN Number 3265
Formula Weight 128.57
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Ethanesulfonyl Chloride
EINECSNumber (2)-3915
RTECSNumber KI8050000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.