Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanethiol, 99+%

Catalog No. AC117860025
Change view
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117860025 2.5 L Glass bottle
AC117860500 50 mL Glass bottle
AC117865000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC117860025 Supplier Thermo Scientific Chemicals Supplier No. 117860025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 75-08-1 | C2H6S | 62.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-08-1
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00004887
InChI Key DNJIEGIFACGWOD-UHFFFAOYSA-N
Synonym ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan
PubChem CID 6343
IUPAC Name ethanethiol
SMILES CCS

Specifications

Melting Point -148.0°C
Color Colorless
Density 0.8300g/mL
Boiling Point 35.0°C
Flash Point −45°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4296 to 1.4316
Linear Formula C2H5SH
Quantity 2.5 L
Beilstein 01,34
Fieser 12,29
Merck Index 15,3781
Specific Gravity 0.83
Solubility Information Solubility in water: 6.76g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 62.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethanethiol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Extremely flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away
EINECSNumber 200-837-3
RTECSNumber KI9625000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.