accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75-08-1

IUPAC Name: ethanethiol
Synonyms: ethanethiol
Molecular Weight (g/mol): 62.13
Molecular Formula: C2H6S
InChi Key: DNJIEGIFACGWOD-UHFFFAOYSA-N
SMILES: CCS
Boiling Point 
  • (5)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (6)
  • (5)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (8)
  • (1)
  • (1)