Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 200 proof (100%), USP, Decon™ Labs
GREENER_CHOICE

Catalog No. 04355233 Shop All Decon Labs Products
Change view
Click to view available options
Quantity:
0.125 gal.
5 gal.
4 x 1 gal.
55 gal.
24 x 0.125 gal.
Packaging:
Cubitainer with Spigot
Glass Bottle
Metal Container
Plastic Bottle
Poly Bottle
Poly Bottle with Spigot
Poly Drum
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
04-355-233 5 gal. Metal Container
07-678-005 0.125 gal. Glass Bottle
07-678-004 24 x 0.125 gal. Glass Bottle
04-355-222 24 x 0.125 gal. Plastic Bottle
04-355-720 4 x 1 gal. Glass Bottle
04-355-223 4 x 1 gal. Plastic Bottle
22-032-601 5 gal. Cubitainer with Spigot
04-355-224 5 gal. Poly Bottle
22-032-103 5 gal. Poly Bottle with Spigot
04-355-230 55 gal. Metal Container
04-355-600 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 04-355-233 Supplier Decon Laboratories, Inc. Supplier No. 2805M

This product is not available for sale online. Please contact your sales rep or Customer Service at 1-800-766-7000.

May require a federal tax exempt certificate.

Used for research, healthcare, industrial, or production applications as a cleaner, solvent or reagent.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07 g/mol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name Ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Clear
CAS Min % 99.9%
CAS Max % 100%
Density 6.62
Quantity 5 gal.
Packaging Metal Container
Physical Form Liquid
UN Number UN1170
Boiling Point 78.3°C
Percent Purity 95%
Grade BP/EP/FCC/JP/NF/USP
Chemical Name or Material Ethanol, 200 Proof (100%)
Show More Show Less
DOTInformation DOT Class: 3, Packaging II
Recommended Storage Do not store Ethanol at temperature above 120°F
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.