Learn More
Description
Specifications
Specifications
| Melting Point | -114°C |
| Color | Clear |
| CAS Min % | 99.9% |
| CAS Max % | 100% |
| Density | 6.62 |
| Quantity | 4 x 1 gal. |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| UN Number | UN1170 |
| Boiling Point | 78.3°C |
| Show More |
Safety and Handling
| DOTInformation | DOT Class: 3, Packaging II |
| Recommended Storage | Do not store Ethanol at temperature above 120°F |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
