Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol 70%, Vion Biosciences
SCI_ED

Catalog No. S02511 Shop All Aldon Products
Change view
Click to view available options
Quantity:
4 x 1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02511 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Methanol and Methyl isobutyl ketone, which are known to the State of California to cause cancer and birth defects or other reproductive harm.
Catalog No. S02511 Supplier Aldon Corporation Supplier No. VNEE0069CS/4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Methanol and Methyl isobutyl ketone, which are known to the State of California to cause cancer and birth defects or other reproductive harm.

This 70% ethanol is an excellent choice for long-term storage of histology specimens, at just the right concentration to ensure optimal preservation of tissues.

  • This 70% ethanol is an excellent choice for long-term storage of histology specimens, at just the right concentration to ensure optimal preservation of tissues.

Chemical Identifiers

CAS 7732-18-5 , 67-63-1 , 64-17-5 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanol, 70% Aqueous Solution
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name water
SMILES O

Specifications

pH Data not available.
Melting Point -114°C
Color Colorless
CAS Min % 30.00%, 59.50%, 2.80%, 6.30%
CAS Max % 60.60%, 2.90%, 6.30%, 30.00%
Appearance Clear, colorless liquid.
Decomposition Information Heat/Fire
Density 0.7919 to 0.7955 g/mL
Quantity 4 x 1 gal.
Explosion Limit 3.30%
Flash Point 21°C
Identification 70% Ethanol
Ignition Point 383°C
Infrared Spectrum Yes
Odor Mild Characteristic Odor
Packaging 1 Gallon F Style UN Bottle
Physical Form Liquid
Product Line Vion Histology
Specific Gravity 0.7919 to 0.7955 g/mL
Assay Percent Range 59.5 - 60.06%
Vapor Pressure Ca 50 (at 20°C)
Linear Formula C2H5OH
Water 5.00%
UN Number UN1170
Concentration Colorless
Formulation 70% Ethanol
Boiling Point 74°C to 80°C
Solubility Information Soluble in water.
InChI Formula 1S/C2H6O/c1-2-3/h3H,2H2,1H3
Percent Purity 85%
Grade Histological
Concentration or Composition (by Analyte or Components) Colorless
Chemical Name or Material Ethanol
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Causes skin irritation.
Hazard Statement Causes serious eye irritation.
Precautionary Statement May cause drowsiness or dizziness.
ShelfLife 3 Years
DOTInformation UN1170, Ethanol, 3(II), Limited quantity equal to or less than 1 L
EINECSNumber 200-578-6
RTECSNumber KQ6300000
Recommended Storage Store in a cool, dry, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.