Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Ethanol Assay Kit

Catalog No. 89943657 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-943-657 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-943-657 Supplier Abnova™ Supplier No. KA4784
Add to Cart
Add to Cart

Ethanol Assay Kit is a colorimetric determination of ethanol at 565 nm.

  • Suitable Sample: Plasma, saliva, serum, urine
  • Sample Volume: 10 μL
  • Calibration Range: 0.0008 to 0.1%
  • Quality Control Testing: Standard Curve

Specifications

Content And Storage Store the kit at -20°C.
For Use With (Application) Functional Study
Quantity 1 Kit
Type Activity Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.