Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol-d6, for NMR, anhydrous, 99 atom % D

Catalog No. AC300680010
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC300680010 1 mL
AC300680050 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC300680010 Supplier Thermo Scientific Chemicals Supplier No. 300680010
Add to Cart
This item is not returnable. View return policy

CAS: 1516-08-1 | C2H6O | 52.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1516-08-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 52.106
MDL Number MFCD00051020
InChI Key LFQSCWFLJHTTHZ-LIDOUZCJSA-N
Synonym ethanol-d6, 2h6 ethanol, hexadeuteroethanol, 2 h? ethan 2 h ol, ethyl-d5 alcohol-d, ethyl alcohol-d6, ethanol d, ethanol-d6 9ci
PubChem CID 102138
IUPAC Name 1,1,1,2,2-pentadeuterio-2-deuteriooxyethane
SMILES CCO

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 mL
Deuterium (D) 98.5% min.
Density 0.8920g/mL
Linear Formula C2D5OD
Formula Weight 52.12
Grade NMR
Chemical Name or Material Ethanol-d6, For NMR
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.