Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Ethanol (Denatured) (Ethyl Alcohol)
SDP

Catalog No. 14185643 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-643 500 mL
14-185-256 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-185-643 Supplier Medisca Supplier No. 061608
Add to Cart
Add to Cart

For Laboratory/Industrial Use Only

For Laboratory/Industrial Use Only

Specifications

Chemical Name or Material Ethanol (Denatured) (Ethyl Alcohol)
Quantity 500 mL
Packaging Bottle, Amber Glass, Boston Round, 500mL, 38-439
Molecular Weight (g/mol) Unknown
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.