Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, denatured, Spectrophotometric Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AA44014K2
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44014K2 1 L
AA44014AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44014K2 Supplier Thermo Scientific Chemicals Supplier No. 044014K2
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 64-17-5 | C2H6O | 46.069 g/mol

Ethanol, anhydrous, denatured is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -90°C
Color Colorless
Appearance Colorless liquid
Density 0.789 g/cm3
Quantity 1 L
Flash Point 11°C (52°F)
Ignition Point 455°C (851°F)
Odor Alcohol-like
Packaging Resealable ChemSeal™ Bottle under Argon
Physical Form Liquid
Refractive Index 1.361
Assay Percent Range 90%
Vapor Pressure 57 hPa (43mm Hg) at 20°C
UN Number 1987
Beilstein 1718733
Boiling Point 78°C
Solubility Information Miscible with water and most organic solvents.
Formula Weight 46.08
Grade Spectrophotometry
Concentration or Composition (by Analyte or Components) Ethyl alcohol: 90.0%; Methanol: 5.0%; 2-Propanol: 5.0%
Chemical Name or Material Ethanol, denatured
Show More Show Less
Hazard Category H225-H302+H332-H319-H336-H371
Hazard Statement Keep container tightly closed and in a well-ventilated place. Keep away from sources of ignition - No Smoking. Avoid contact with eyes. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P309+P311-P312-P330-P370+P378q-P501c
DOTInformation Hazard Class: 3, Packing Group: II
EINECSNumber 200-578-6
RTECSNumber KQ6300000
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.