Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, for HPLC, denatured
GSA_VA

Catalog No. ac611050040
Change view
Click to view available options
Quantity:
4 L
Packaging:
EcoSafPak™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC611050040 4 L EcoSafPak™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC611050040 Supplier Thermo Scientific Chemicals Supplier No. 611050040
Only null left
Add to Cart

CAS: 64-17-5 | C2H6O | 46.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point 114°C
Absorbance 0.04 max. at 254nm, 0.35 max. at 220nm, 0.65 max. at 210nm, 1 max. at 205nm
Density 1.24 (Air = 1.00)
Quantity 4 L
Flash Point 12.8°C (55°F)
Identification Pass Test
Packaging EcoSafPak™
Refractive Index 1.3580 - 1.3610
Residue after Evaporation 5ppm max.
Titratable Acid 0.0003 meq/g max.
Linear Formula C2H5OH
Titratable Base 0.0002 meq/g max.
Water 0.10% max.
Merck Index 14, 3761
Boiling Point 78°C
Formula Weight 46.06
Grade HPLC
Chemical Name or Material Ethanol, For HPLC, Denatured
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause damage to organs.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.