Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Ethanol Sensor
SDPSCI_ED

Catalog No. S16146ND Shop All Vernier Software Products
Change view
Click to view available options
Range:
0 to 3%
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S16146ND 0 to 3%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16146ND Supplier Vernier™ Supplier No. ETHBTA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Ethanol Sensor, Spare cap assembly, Tape for Ethanol Sensor, #6 Stopper for Ethanol Sensor, 250 mL Nalgene Bottle w/Lid

Requires

Requires an interface and software, sold separately

The Ethanol Sensor can be used to measure the concentration of ethanol in air above an aqueous sample.

Range: 0% to 3% Typical Resolution: Concentration Resolution 2-3% 0.02% 1-2% 0.01% 0-1% 0.001% Response Time: 95% of full scale in 60 seconds.

Warranty and Services

5-year limited warranty

Specifications

Grade High School and College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Range 0 to 3%
Resolution 0.02% at 2 to 3% concentration, 0.01% at 1 to 2% concentration, 0.001% at 0 to 1% concentration
Response Time 95% of full scale in 60 seconds
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.