Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol for Spectroscopy Uvasol™ MilliporeSigma™

Catalog No. M1009800500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1009800500 500 mL Glass Bottle
M1009802500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1009800500 Supplier MilliporeSigma Supplier No. 1.00980.0500
Add to Cart
Add to Cart

For spectroscopy Uvasol™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

pH 7 (at 10g/L 20°C)
Melting Point -114.5°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 500 mL
Flash Point 12°C (53.6°F) Closed cup
Ignition Point 425°C (797.9°F)
Odor Alcohol-like
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 59 hPa (20°C)
UN Number 1170
Solubility Information Soluble in water at 20°C, completely miscible
Boiling Point 78.3°C
Formula Weight 46.07
Chemical Name or Material Ethanol
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Recommended Storage Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.