Learn More
Medchemexpress LLC Ethephon-d4 | 1020719-29-2 | >98.0% | 148.52 g/mol | C2H2D4ClO3P | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW131845S10MG
Encompass_Preferred
Ethephon-d4 is the deuterium-labeled analogue of ethephon intended for analytical and research use. It functions as a tracer and internal standard for quantitative methods and is suitable for NMR, GC-MS, and LC-MS applications.
  • Deuterium-labeled internal standard suitable for quantitative analysis.
  • High deuterium enrichment (≈99 atom % D) with chemical purity ≥98%.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Stable when stored sealed at 4°C; in solution: -80°C (up to 6 months) or -20°C (≈1 month).
  • Supplied in small calibrated quantities for analytical use.
Catalog No. 50-002-83319