Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethox Titrating Reagent, Reagents
Ethox Titrating Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc C84010NE1C
Quantity | 1 L |
---|
Chemical Identifiers
7732-18-5, 207683-22-5 | |
18.02 | |
water |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
O |
Specifications
Colorless | |
100°C | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
1 | |
Non-sterile | |
Ethox Titrating Reagent |
7732-18-5, 207683-22-5 | |
H2O | |
O | |
18.02 | |
Glass Bottle | |
1 L | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction