Learn More
Sigma Organic Chemistry Ethylene glycol monophenyl ether | 1L | 122-99-6 | MFCD00002857 | >=90%
Supplier: Sigma Organic Chemistry 4843181L

Ethylene glycol monophenyl ether, 1L
About This Item:
Synonym(s): DOWANOL(R) EPh; 2-Phenoxyethanol; Phenylglycol
Linear Formula: C6H5OCH2CH2OH
Molecular Weight: 138.16
CAS Number: 122-99-6
MDL Number: MFCD00002857
UNSPSC Code: 12162002
Purity: >=90%
Boiling Point: 247 C (lit.)
Melting Point: 11-13 C (lit.)
Density: 1.102 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.537 (lit.)
Storage: room temp
EINECS Number: 204-589-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.