Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1,5-Dimethylpyrazole-3-carboxylate 98.0+%, TCI America™
SDP

Catalog No. E10021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10021G 1 g
E1002200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10021G Supplier TCI America Supplier No. E10021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5744-51-4
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00085071
InChI Key OJPXVXXMBWKEAT-UHFFFAOYSA-N
PubChem CID 138577
IUPAC Name ethyl 1,5-dimethyl-1H-pyrazole-3-carboxylate
SMILES CCOC(=O)C1=NN(C)C(C)=C1

Specifications

Melting Point 45°C
Color Yellow
Boiling Point 119°C
Quantity 1 g
Synonym ethyl 1,5-dimethyl-1h-pyrazole-3-carboxylate, 1,5-dimethyl-3-ethoxycarbonylpyrazole, ethyl1,5-dimethyl-1h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, 1,5-dimethyl-, ethyl ester, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid ethyl ester, 1,5-dimethylpyrazole-3-carboxylic acid ethyl ester, 1,5-dimethyl-1 h-pyrazole-3-carboxylic acid ethyl ester, acmc-209lya, maybridge1_008890, ksc606q7n
Formula Weight 168.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1,5-Dimethylpyrazole-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.