Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-(2-amino-5-nitrophenyl)-4-methylthiazole-5-carboxylate, 97%, Thermo Scientific™

Catalog No. AAH6699303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6699303 1 g
AAH66993MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6699303 Supplier Thermo Scientific Chemicals Supplier No. H6699303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41450-33-3
Molecular Formula C13H13N3O4S
Molecular Weight (g/mol) 307.324
InChI Key ASHVHIIHZOANMU-UHFFFAOYSA-N
Synonym ethyl 2-2-amino-5-nitrophenyl-4-methylthiazole-5-carboxylate, ethyl 2-2-amino-5-nitrophenyl-4-methyl-1,3-thiazole-5-carboxylate
PubChem CID 86277503
IUPAC Name ethyl 2-(2-amino-5-nitrophenyl)-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)C2=C(C=CC(=C2)[N+](=O)[O-])N)C

Specifications

Melting Point 238°C to 242°C
Color Yellow
Quantity 1 g
Formula Weight 307.33
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material Ethyl 2-(2-amino-5-nitrophenyl)-4-methylthiazole-5-carboxylate
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.