Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-(2-chloroacetamido)-5-phenylthiophene-3-carboxylate

Catalog No. OTV0008711G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0008711G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0008711G Supplier Sigma Aldrich Supplier No. OTV0008711G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl valerate is a methyl ester of valeric acid, generally used as a fragrance in the manufacture of soaps and laundry detergents. it is also used as a plasticizer and surrogate component for
  • Biodiesel.methyl valerate can be used:
  • In the preparation of lankacidin c 8-valerate
  • As a reactant to synthesize bis(2,2,2-trifluoroethyl) 2-oxoalkylphosphonates
  • As a substrate in the fluorination reactions of unactivated c(sp3)h bonds using decatungstate photocatalyst and n-fluorobenzenesulfonimide

Specifications

Molecular Formula C15H14ClNO3S
Linear Formula C15H14ClNO3S
MDL Number MFCD02350907
Quantity 100 mL
Molecular Weight (g/mol) 323.79
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.