Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-(2-Methyl-1,3-dioxolan-2-yl)acetate 98.0+%, TCI America™
SDP

Catalog No. E0929100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0929100G 100 g
E092925G 25 g
E0929500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0929100G Supplier TCI America Supplier No. E0929100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6413-10-1
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00152488
InChI Key XWEOGMYZFCHQNT-UHFFFAOYSA-N
Synonym 2-(2-Methyl-1,3-dioxolan-2-yl)acetic Acid Ethyl Ester
PubChem CID 80865
IUPAC Name ethyl 2-(2-methyl-1,3-dioxolan-2-yl)acetate
SMILES CCOC(=O)CC1(OCCO1)C

Specifications

Color Colorless
Boiling Point 98°C
Quantity 100 g
Formula Weight 174.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-(2-Methyl-1,3-dioxolan-2-yl)acetate
EINECSNumber (5)-1178
RTECSNumber JH6762500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.