Learn More
Sigma Organic Chemistry Ethyl 2- 2-tolyl thiazo | 1G | MFCD28166210
Supplier: Sigma Organic Chemistry BOG001481G

Ethyl 2- 2-tolyl thiazo , 1G
About this Item:
MDL #: MFCD28166210
Molecular Weight: 323.41
UNSPSC Code: 12352200
Chemical Formula: C19H17NO2S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.