Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2,3,4,5-Tetrafluorobenzoate 98.0+%, TCI America™
SDP

Catalog No. T2094100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2094100G 100 g
T209425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. T2094100G Supplier TCI America Supplier No. T2094100G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 122894-73-9
Molecular Formula C9H6F4O2
Molecular Weight (g/mol) 222.139
MDL Number MFCD02094158
InChI Key SBHMXBYMQZRRLC-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorobenzoic acid ethyl ester, 2,3,4,5-tetrafluoro-benzoic acid ethyl ester, pubchem4072, acmc-1c5fu, ethyl2,3,4,5-tetrafluorobenzoate, 2,3,4,5-tetrafluoro-benzoicacidethylester, ethyl 2,3,4,5-tetrakis fluoranyl benzoate
PubChem CID 2774395
IUPAC Name ethyl 2,3,4,5-tetrafluorobenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1F)F)F)F

Specifications

Color Colorless
Boiling Point 88°C
Quantity 100 g
Formula Weight 222.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2,3,4,5-Tetrafluorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.