Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2,5-Dichlorobenzoate 98.0+%, TCI America™
SDP

Catalog No. D256625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D256625G 25 g
D25665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. D256625G Supplier TCI America Supplier No. D256625G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Chemical Identifiers

CAS 35112-27-7
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.061
MDL Number MFCD00013636
InChI Key JSZYWIKNIZKJAN-UHFFFAOYSA-N
PubChem CID 123447
IUPAC Name ethyl 2,5-dichlorobenzoate
SMILES CCOC(=O)C1=C(C=CC(=C1)Cl)Cl

Specifications

Color Yellow
Quantity 25 g
Synonym 2,5-dichlorobenzoic acid ethyl ester, ethyl-2,5-dichlorobenzoate, pubchem3706, acmc-1ajde, rarechem al bi 0427, jszywiknizkjan-uhfffaoysa, ethyl 2,5-bis chloranyl benzoate, labotest-bb lt00454292, benzoic acid,2,5-dichloro-, ethyl ester, benzoic acid, 2,5-dichloro-, ethyl ester
Formula Weight 219.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2,5-Dichlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.