Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, 95%

Catalog No. AC446640010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446640010 1 g
AC446640050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446640010 Supplier Thermo Scientific Chemicals Supplier No. 446640010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 193140-71-5 | C5H9ClF3NO2 | 207.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193140-71-5
Molecular Formula C5H9ClF3NO2
Molecular Weight (g/mol) 207.58
MDL Number MFCD14525479
InChI Key MNFISCBCWBUTTQ-UHFFFAOYNA-N
Synonym ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride, 2-amino-3,3,3-trifluoro-propionic acid ethyl ester hydrochloride, ethyl 3,3,3-trifluoroalaninate, ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, ethyl 3,3,3-trifluoroalaninate-hydrogen chloride 1/1
PubChem CID 45790935
IUPAC Name ethyl 2-amino-3,3,3-trifluoropropanoate;hydrochloride
SMILES Cl.CCOC(=O)C(N)C(F)(F)F

Specifications

Melting Point 218°C to 220°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94.0 to max. 106% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 207.58
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 2-amino-3, 3, 3-trifluoropropionate hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact l

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.