Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 99%, Thermo Scientific™

Catalog No. AC251280250
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC251280250 25 g
AC251280100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC251280250 Supplier Thermo Scientific Chemicals Supplier No. 251280250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4506-71-2
InChI Key CDYVTVLXEWMCHU-UHFFFAOYSA-N
PubChem CID 78262
IUPAC Name ethyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC2=C1CCCC2)N

Specifications

Melting Point 114°C to 117°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C11H15NO2S
Quantity 25 g
Synonym ethyl 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid ethyl ester, 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid ethyl ester, benzo b thiophene-3-carboxylic acid, 2-amino-4,5,6,7-tetrahydro-, ethyl ester, ethyl2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate, methylaminotetrahydrobenzothiophenecarboxylate, 2-amino-3-carbethoxy-4,5-tetramethylenethiophene, ethyl 2-amino-4,5,6,7-tetrahydrobenzothiophene-3-carboxylate, ethyl-2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylate
Molecular Weight (g/mol) 225.306
Formula Weight 225.31
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-823-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.