Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-(bromomethyl)acrylate

Catalog No. 4252221G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-522-21G 1 g
42-522-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-522-21G Supplier Sigma Aldrich Supplier No. 4252221G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl 2-(bromomethyl)acrylate is an allylic alkylating reagent.It can be employed as an electrophile for various organometallic compounds. The organometallic derivatives of the compound can be used for the synthesis ofα-methylene lactones and lactams. It can also be used for the polymerization of functionalized acrylic monomers.

Specifications

CAS 17435-72-2
Density 1.398 g/mL (at 25°C)
Boiling Point 38°C (0.8 mmHg) (literature)
Molecular Formula C6H9BrO2
Refractive Index n20/D 1.479 (literature)
Linear Formula CH2=C(CH2Br)CO2CH2CH3
Quantity 1 g
MDL Number MFCD00031518
Synonym 2-(Bromomethyl)acrylic acid ethyl ester; Ethyl alpha-(bromomethyl)acrylate; Ethyl 2-(bromomethyl)-2-propenoate; Ethyl 2-(bromomethyl)propenoate
Molecular Weight (g/mol) 193.04
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.