Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-(bromomethyl)acrylate

Catalog No. 4252225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-522-25G 5 g
42-522-21G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-522-25G Supplier Sigma Aldrich Supplier No. 4252225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl 2-(bromomethyl)acrylate is an allylic alkylating reagent.It can be employed as an electrophile for various organometallic compounds. The organometallic derivatives of the compound can be used for the synthesis ofα-methylene lactones and lactams. It can also be used for the polymerization of functionalized acrylic monomers.

Specifications

CAS 17435-72-2
Density 1.398 g/mL (at 25°C)
Boiling Point 38°C (0.8 mmHg) (literature)
Molecular Formula C6H9BrO2
Refractive Index n20/D 1.479 (literature)
Linear Formula CH2=C(CH2Br)CO2CH2CH3
Quantity 5 g
MDL Number MFCD00031518
Synonym 2-(Bromomethyl)acrylic acid ethyl ester; Ethyl alpha-(bromomethyl)acrylate; Ethyl 2-(bromomethyl)-2-propenoate; Ethyl 2-(bromomethyl)propenoate
Molecular Weight (g/mol) 193.04
Percent Purity 98%
Biological Activity Respiratory system
Afficher plus de résultats Afficher moins de résultats
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.