Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-chloropropionate, 97%, Thermo Scientific™

Catalog No. AAL1341030
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1341030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1341030 Supplier Thermo Scientific Chemicals Supplier No. L1341030

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-13-7
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00000864
InChI Key JEAVBVKAYUCPAQ-UHFFFAOYNA-N
Synonym ethyl 2-chloropropionate, 2-chloropropionic acid ethyl ester, propanoic acid, 2-chloro-, ethyl ester, ethyl-2-chloropropionate, 2-chloropropionic acid, ethyl ester, ethyl alpha-chloropropionate, ethyl .alpha.-chloropropionate, propionic acid, 2-chloro-, ethyl ester, +-ethyl 2-chloropropionate, ethyl ?-chloropropionate
PubChem CID 10807
IUPAC Name ethyl 2-chloropropanoate
SMILES CCOC(=O)C(C)Cl

Specifications

Density 1.072
Boiling Point 146°C to 149°C
Flash Point 38°C (100°F)
Odor Pungent
Refractive Index 1.417
Quantity 250 g
UN Number UN2935
Beilstein 773906
Merck Index 14,3785
Formula Weight 136.58
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloropropionate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYL 2-CHLOROPROPIONATE
EINECSNumber 208-610-0
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.