Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Cyano-3,3-diphenylacrylate 98.0+%, TCI America™
SDP

Catalog No. C096225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C096225G 25 g
C0962500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C096225G Supplier TCI America Supplier No. C096225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5232-99-5
Molecular Formula C18H15NO2
Molecular Weight (g/mol) 277.323
MDL Number MFCD00027364
InChI Key IAJNXBNRYMEYAZ-UHFFFAOYSA-N
Synonym 2-Cyano-3,3-diphenylacrylic Acid Ethyl Ester
PubChem CID 243274
IUPAC Name ethyl 2-cyano-3,3-diphenylprop-2-enoate
SMILES CCOC(=O)C(=C(C1=CC=CC=C1)C2=CC=CC=C2)C#N

Specifications

Melting Point 98°C
Color White-Yellow
Boiling Point 200°C
Quantity 25 g
Formula Weight 277.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Cyano-3,3-diphenylacrylate
EINECSNumber (2)-2792&(4)-0262
RTECSNumber AT6200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.