Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Formylpropionate 97.0+%, TCI America™
SDP

Catalog No. F04111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04111G 1 g
F04115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04111G Supplier TCI America Supplier No. F04111G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27772-62-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00044012
InChI Key VVCYNVCCODBCOE-UHFFFAOYSA-N
Synonym 2-Formylpropionic Acid Ethyl Ester
PubChem CID 10898741
IUPAC Name ethyl 2-methyl-3-oxopropanoate
SMILES CCOC(=O)C(C)C=O

Specifications

Color Yellow
Quantity 1 g
UN Number 1993
Formula Weight 130.14
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Formylpropionate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.