Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Hydroxybutyrate 95.0+%, TCI America™
SDP

Catalog No. H022925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H022925G 25 g
H02295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H022925G Supplier TCI America Supplier No. H022925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52089-54-0
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00014407
InChI Key KWWOQRSLYPHAMK-UHFFFAOYNA-N
Synonym 2-Hydroxybutyric Acid Ethyl Ester
PubChem CID 521365
IUPAC Name ethyl 2-hydroxybutanoate
SMILES CCOC(=O)C(O)CC

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 g
Formula Weight 132.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Hydroxybutyrate
EINECSNumber (2)-1430
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.