Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-mercaptoacetate, 99%

Catalog No. AC118670025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118670025 2.5 kg Glass bottle
AC118671000 100 g Glass bottle
AC118675000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC118670025 Supplier Thermo Scientific Chemicals Supplier No. 118670025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 623-51-8 | C4H8O2S | 120.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-51-8
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00004874
InChI Key PVBRSNZAOAJRKO-UHFFFAOYSA-N
Synonym ethyl thioglycolate, ethyl mercaptoacetate, ethyl 2-mercaptoacetate, mercaptoacetic acid ethyl ester, ethyl thioglycollate, acetic acid, mercapto-, ethyl ester, thioglycolic acid ethyl ester, ethyl mercaptoacetic acid, usaf ek-2070, acetic acid, 2-mercapto-, ethyl ester
PubChem CID 12185
IUPAC Name ethyl 2-sulfanylacetate
SMILES CCOC(=O)CS

Specifications

Melting Point -80°C
Color Colorless
Density 1.0900g/mL
Boiling Point 54°C (12.0 mmHg)
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4560 to 1.459
Linear Formula HSCH2CO2C2H5
Quantity 2.5 kg
Beilstein 03,255
Specific Gravity 1.09
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone
Viscosity 1.2 mPa.s (20°C)
Formula Weight 120.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl 2-mercaptoacetate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-800-3
RTECSNumber AI6650000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.