Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Methylvalerate 98.0+%, TCI America™
SDP

Catalog No. E080125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E080125ML 25 mL
E0801500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E080125ML Supplier TCI America Supplier No. E080125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39255-32-8
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00036564
InChI Key HZPKNSYIDSNZKW-UHFFFAOYNA-N
Synonym ethyl 2-methylvalerate, manzanate, pentanoic acid, 2-methyl-, ethyl ester, melon valerate, ethyl alpha-methylvalerate, ethyl .alpha.-methylvalerate, fema no. 3488, ethyl, a-methylvalerate, acmc-1ahki, ethyl 2-methyl pentanoate
PubChem CID 62902
IUPAC Name ethyl 2-methylpentanoate
SMILES CCCC(C)C(=O)OCC

Specifications

Color Colorless
Boiling Point 153°C
Quantity 25 mL
UN Number 3272
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Methylvalerate
EINECSNumber (2)-0782
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.