Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Amino-4,4,4-trifluorocrotonate 97.0+%, TCI America™
SDP

Catalog No. A147525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A147525G 25 g
A14755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A147525G Supplier TCI America Supplier No. A147525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 372-29-2
Molecular Formula C6H8F3NO2
Molecular Weight (g/mol) 183.13
MDL Number MFCD00068195
InChI Key NXVKRKUGIINGHD-ONEGZZNKSA-N
PubChem CID 10899311
IUPAC Name ethyl (E)-3-amino-4,4,4-trifluorobut-2-enoate
SMILES CCOC(=O)C=C(C(F)(F)F)N

Specifications

Color White-Yellow
Boiling Point 83°C
Quantity 25 g
Synonym ethyl 3-amino-4,4,4-trifluorocrotonate, ethyl 3-amino-4,4,4-trifluorobut-2-enoate, ethyl 2z-3-amino-4,4,4-trifluorobut-2-enoate, 3-amino-4,4,4-trifluorocrotonic acid ethyl ester, ethyl-3-amino-4,4,4-trifluorocrotonate, ethyl 3-amino-4,4,4-trifluoro-2-butenoate, ethyl z-3-amino-4,4,4-trifluorobut-2-enoate, ethyl z-3-amino-4,4,4-trifluoro-but-2-enoate, hcvdpjfpdddldlbdlrfuulu@bh, ethyl3-amino-4,4,4-trifluorocrotonate
Formula Weight 183.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 3-Amino-4,4,4-trifluorocrotonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.