Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Carbamimidoylamino-4-methylbenzoate Nitrate 97.0+%, TCI America™
SDP

Catalog No. E10791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10791G 1 g
E10795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10791G Supplier TCI America Supplier No. E10791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 641569-96-2
Molecular Formula C11H16N4O5
Molecular Weight (g/mol) 284.27
MDL Number MFCD11100711
InChI Key YQMZYKJPGMVZJL-UHFFFAOYSA-N
Synonym 3-Carbamimidoylamino-4-methylbenzoic Acid Ethyl Ester Nitrate, Ethyl 3-Carbamimidoylamino-p-toluate Nitrate, 3-Carbamimidoylamino-p-toluic Acid Ethyl Ester Nitrate
PubChem CID 44181764
IUPAC Name ethyl 3-[(diaminomethylidene)amino]-4-methylbenzoate; nitric acid
SMILES O[N+]([O-])=O.CCOC(=O)C1=CC(N=C(N)N)=C(C)C=C1

Specifications

Melting Point 199°C
Color White
Quantity 1 g
Formula Weight 284.27
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Carbamimidoylamino-4-methylbenzoate Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.